Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Texas Red TCO
Texas Red TCO fluorescent probe
Texas Red TCO fluorescent probe
AATB899
:
AAT Bioquest
:
Texas Red TCO fluorescent probe
AATB899
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
815.01 g/mol
:
5,II
:
GHS07