Fluorescamine
Plastic bottle, 25 mg
Catalog # AATB820
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Fluorescamine is a popular amine-fluorogenic reagent for determining protein concentrations in solutions and on gels.
Synonyms: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
- CAS Number:38183-12-9
- Formula:C₁₇H₁₀O₄
- MDL Number:MFCD00005928
- Molecular Weight:278.26 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



