Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor®
iFluor® 546, Styramide "Superior Replacement for Alexa Fluor 546 tyramide"
iFluor® 546, Styramide "Superior Replacement for Alexa Fluor 546 tyramide"
iFluor® 546, Styramide "Superior Replacement for Alexa Fluor 546 tyramide"
Catalog # AATB45025
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
iFluor® 546, Styramide "Superior Replacement for Alexa Fluor 546 tyramide"
Catalog # AATB45025
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
100
Seq:
0100
Specifications
About this item
Product Details
:
Product Details