Order Entry
Germany
ContactUsLinkComponent
5-TAMRA C6 maleimide ≥95%
5-TAMRA C6 maleimide ≥95%
Catalog # AATB424
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
5-TAMRA C6 maleimide ≥95%
Catalog # AATB424
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :623.67 g/mol