
PG-cholesterylamine for cholesterol measurement
Artikelnr. AATB36700
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:1043.73 g/mol
ADR:5,II
:GHS07
CAS Number:1264272-41-4