Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulpho-Cyanine 5 NHS ester
Sulpho-Cyanine 5 NHS ester ≥90%
Sulpho-Cyanine 5 NHS ester ≥90%
Sulpho-Cyanine 5 NHS ester ≥90%
Supplier:
AAT BIOQUEST
Certificates
Expand image
Sulpho-Cyanine 5 NHS ester ≥90%
Supplier:
AAT BIOQUEST
About this item
CAS Number:
1448789-45-4
: