0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
PE-iFluor™ 700 Tandem
PE-iFluor™ 700 Tandem, ReadiUse™ fluorescent probe, preactivated
PE-iFluor™ 700 Tandem, ReadiUse™ fluorescent probe, preactivated
AATB2585
:
AAT Bioquest
:
PE-iFluor™ 700 Tandem, ReadiUse™ fluorescent probe, preactivated
AATB2585
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
5,II
:
GHS07