Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cy3DIGE NHS ester
Cy3DIGE NHS ester ≥90%
Cy3DIGE NHS ester ≥90%
Cy3DIGE NHS ester ≥90%
AATB25303
:
AAT BIOQUEST
:
Cy3DIGE NHS ester ≥90%
AATB25303
:
AAT BIOQUEST
:
Pk:
100 Nmo
Seq:
9040
:
:
709.67 g/mol