Order Entry
Germany
ContactUsLinkComponent
Indo-1 pentasodium salt ratiometric calcium indicator
Indo-1 pentasodium salt ratiometric calcium indicator
Catalog # AATB21044
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Indo-1 pentasodium salt ratiometric calcium indicator
Catalog # AATB21044
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :759.5 g/mol
ADR:5,II
 :GHS07
 :132319-56-3
Formula:C₃₂H₂₆N₃Na₅O₁₂