Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fura 2-AM
Fura 2-AM ≥95%
Fura 2-AM ≥95%
Irritant
Fura 2-AM ≥95%
Supplier:
AAT BIOQUEST
Certificates
Expand image
Irritant
Fura 2-AM ≥95%
Supplier:
AAT BIOQUEST
Certificates
About this item
CAS Number:
108964-32-5
:
Formula:
C₄₄H₄₇N₃O₂₄
MDL Number:
MFCD00036976
ADR:
5,II
:
1001.857 g/mol
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
Documentation