BAPTA-AM ≥95%
Plastic bottle, 25 mg
Catalog # AATB21001
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
BAPTA AM is cell-permeable version of BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid), a cell-permeable calcium chelator.
Synonyms: Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM ester
- CAS Number:126150-97-8
- Formula:C₃₄H₄₀N₂O₁₈
- MDL Number:MFCD00036696
- Molecular Weight:764.69 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



