Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fura-8FF™ pentapotassium salt
Fura-8FF™ pentapotassium salt
Fura-8FF™ pentapotassium salt
Fura-8FF™ pentapotassium salt
Catalog # AATB20621
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Fura-8FF™ pentapotassium salt
Catalog # AATB20621
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
500 µG
Seq:
1418
Specifications
About this item
Product Details
:
:
837.97 g/mol
ADR:
5,II
:
GHS07
Product Details