Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluo-4 AM
Fluo-4 AM ≥95%, Ultra Pure Grade
Fluo-4 AM ≥95%, Ultra Pure Grade
Fluo-4 AM ≥95%, Ultra Pure Grade
Supplier:
AAT BIOQUEST
Certificates
Expand image
Fluo-4 AM ≥95%, Ultra Pure Grade
Supplier:
AAT BIOQUEST
About this item
CAS Number:
273221-67-3
:
Formula:
C₅₁H₅₀F₂N₂O₂₃
:
1096.95 g/mol
ADR:
5,II
:
GHS07