0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
ICG-OSu
ICG-OSu ≥90%
ICG-OSu ≥90%
Catalog # AATB182
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
ICG-OSu ≥90%
Catalog # AATB182
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
828.03 g/mol
CAS Number:
1622335-40-3
Product Details