Order Entry
Germany
ContactUsLinkComponent
Hoechst 34580 trihydrochloride ≥95%
Irritant
Hoechst 34580 trihydrochloride ≥95%
Catalog # AATB17537
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Hoechst 34580 trihydrochloride ≥95%
Catalog # AATB17537
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :560.95 g/mol
ADR:5,II
 :GHS07
CAS Number:911004-45-0
 :23555-00-2
Formula:C₂₇H₂₉N₇ . 3HCl