Order Entry
Germany
ContactUsLinkComponent
7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP
7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP
Catalog # AATB17092
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP
Catalog # AATB17092
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 Nmo
  • Seq:
    9030

Specifications

About this item

 :
 :614.3 g/mol
ADR:5,II
 :GHS07
CAS Number:666847-81-0
Formula:C15H21N8O13P3