0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
DEAC-dUTP
DEAC-dUTP 1 mM in TRIS buffer (pH 7.5)
DEAC-dUTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17024
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
DEAC-dUTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17024
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
25 Nmo
Seq:
0311
Specifications
About this item
Product Details
:
:
945.58 g/mol
ADR:
5,II
:
GHS07
Product Details