XFD488 alkyne fluorescent probe
Plastic bottle, 1 mg
Catalog # AATB1700
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
The fluorescent dye alkynes react with azides via a copper-catalyzed click reaction (CuAAC).
- pH-insensitive
- High photostability
- Brighter signal
- Molecular Weight:773.92 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



