Order Entry
Germany
ContactUsLinkComponent
1,2-Diamino-9,10-anthraquinone ≥95%
Irritant
1,2-Diamino-9,10-anthraquinone ≥95%
Catalog # AATB15220
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
1,2-Diamino-9,10-anthraquinone ≥95%
Catalog # AATB15220
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
ADR:5,II
 :238.241 g/mol
 :GHS07
CAS Number:1758-68-5
Formula:C₁₄H₁₀N₂O₂
MDL Number:MFCD00001219