Dihydrofluorescein diacetate ≥95%
Catalog # AATB15203
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:418.401 g/mol
ADR:5,II
:GHS07
CAS Number:35340-49-9
Formula:C₂₄H₁₈O₇
MDL Number:MFCD00005056