Order Entry
Germany
ContactUsLinkComponent
Cyanine 3 bissuccinimidyl ester ≥90% [equivalent to Cy3® bisNHS ester]
Cyanine 3 bissuccinimidyl ester ≥90% [equivalent to Cy3® bisNHS ester]
Catalog # AATB138
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 3 bissuccinimidyl ester ≥90% [equivalent to Cy3® bisNHS ester]
Catalog # AATB138
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1012.2 g/mol