Order Entry
Germany
Orders LinkContactUsLinkComponent
(Ac-IETD)2-R110

(Ac-IETD)2-R110

Plastic bottle, 1 mg
Catalog # AATB13431
Irritant
Supplier:   AAT Bioquest
(Ac-IETD)2-R110
Catalog # AATB13431
Supplier:  AAT Bioquest
CAS Number:  
(Ac-IETD)2-R110
(Ac-IETD)2-R110

Plastic bottle, 1 mg

CAS Number:  
Price
156,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Since highly purified rhodamine 110 (R110)-derived substrates are locked in the lactone configuration they are colorless and non-fluorescent.

  • Caspase detection
  • Bright signal
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.