Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide solution 50 mM stabilised, ReadiUse™
Hydrogen peroxide solution 50 mM stabilised, ReadiUse™
Catalog # AATB11004
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Hydrogen peroxide solution 50 mM stabilised, ReadiUse™
Catalog # AATB11004
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
ADR:5,II
 :34.01 g/mol
 :GHS07
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333