0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
iFluor®
iFluor® 450, Succinimidyl Ester
iFluor® 450, Succinimidyl Ester
iFluor® 450, Succinimidyl Ester
Supplier:
AAT BIOQUEST
Certificates
Expand image
iFluor® 450, Succinimidyl Ester
Supplier:
AAT BIOQUEST
About this item
:
Storage Temperature:
Freezer