Order Entry
Germany
ContactUsLinkComponent
Native Casein (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Native Casein (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Catalog # AATB13440
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Casein (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Catalog # AATB13440
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    5 mg
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/peptide type:
    Native
  • Source:
    Eukaryotic cells
  • Protein synonyms:
    protease substrate, milk protein

Specifications

About this item

Casein is considered to be a generic substrate for a broad spectrum of proteases.

  • Excellent fluorescence intensity change
  • Signal directly proportional to protease activity
  • Bright signal
Caution: For research use only.