MegaWox™ polyHRP-Protein A Conjugate
Catalog # AATB11041
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:1 mg
- Description:MegaWox™ polyHRP-Protein A Conjugate
Specifications
About this item
Protein A is a 42 kDa protein found in the cell wall of Staphylococcus aureus.
- No unconjugated hrp
- Highly purified
- Compatible with all major substrates
- Multiple applications
- High sensitivity
Caution: For research use only.