Nuclear Red™ LCS1
Catalog # AATB17542
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:0,5 mL
- Description:Nuclear Red™ LCS1
- Purity:95%
Specifications
About this item
Our Nuclear Red™ LCS1 is a fluorogenic, DNA-selective and cell-permeant dye for analysing DNA content in living cells.
- Cell-impermeable
- Useful for multicolour imaging
- Bright fluorescence
5 mM DMSO solution.
Caution: For research use only.