Nuclear Green™ DCS1
Catalog # AATB17550
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:0,5 mL
- Description:Nuclear Green™ DCS1
- Purity:90%
Specifications
About this item
Our Nuclear Green™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analysing DNA content in dead, fixed or apoptotic cells.
- Cell-impermeable
- Useful for multicolour imaging
- Bright fluorescence
5 mM DMSO solution.
Caution: For research use only.