Order Entry
Germany
ContactUsLinkComponent
Native Human Biocytin (from Human), Cal-520®
Native Human Biocytin (from Human), Cal-520®
Catalog # AATB20606
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Native Human Biocytin (from Human), Cal-520®
Catalog # AATB20606
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    250 µG
  • Conjugation:
    Cal-520®
  • Protein/peptide type:
    Native
  • Species:
    Human
  • Protein synonyms:
    Biotinyl
  • Protein/peptide name:
    Biocytin

Specifications

About this item

Biotin and biocytin bind with high affinity to both avidins and streptavidins.

  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant
Caution: For research use only.