Order Entry
Germany
ContactUsLinkComponent
Sunnyvale Red™ SE *Superior 6-ROX Replacement*
Sunnyvale Red™ SE *Superior 6-ROX Replacement*
Catalog # AATB394
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Sunnyvale Red™ SE *Superior 6-ROX Replacement*
Catalog # AATB394
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Description:
    Sunnyvale Red™ dye
  • Purity:
    90%

Specifications

About this item

Although ROX dyes have strong fluorescence they are notoriously unstable.

  • Multiple Bioconjugation Applications
  • 90% Purity
Caution: For research use only.