Texas Red® Maleimide, Single Isomer
Catalog # AATB483
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:5 mg
- Description:Texas Red® maleimide
- Purity:95%
Specifications
About this item
Texas Red® maleimide is an excellent thiol-reactive fluorescent reagent for labelling thiol-containing biological molecules (such as peptides, proteins and thiol-modified oligonucleotides).
- Identical spectra to Texas Red
- High Stability
Caution: For research use only.