Order Entry
Germany
ContactUsLinkComponent
 
ReadiLink™ Protein Conjugation Stop Buffer (1 ml)
Catalog # AATB5400
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ Protein Conjugation Stop Buffer (1 ml)
Catalog # AATB5400
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    ReadiLink™ protein conjugation stop buffer

Specifications

About this item

ReadiLink™ Protein Conjugation Stop Buffer is used to stop the protein conjugation reactions that use succinimidyl ester, sulfonyl chloride or isothiocyanate.

  • Usable at room temperature
  • Stops all major protein conjugation reactions
  • Simple protocol
Caution: For research use only.