Order Entry
Germany
ContactUsLinkComponent
Ac-DEVD-AFC
Ac-DEVD-AFC
Catalog # AATB13401
Supplier:  AAT Bioquest
CAS Number:  
undefined
Ac-DEVD-AFC
Catalog # AATB13401
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • CAS No.:
    201608-14-2
  • Protein/peptide name:
    Ac-DEVD-AFC
  • Purity:
    90%

Specifications

About this item

Ac-DEVD-AFC is a fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.

  • Hazard Class/ Packing Group: 5, II
  • GHS symbol: GHS07
  • Signal Word: Warning