Order Entry
Germany
ContactUsLinkComponent
 
MycoLight™ Green JJ99 *5 mM in DMSO*
Catalog # AATB24001
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MycoLight™ Green JJ99 *5 mM in DMSO*
Catalog # AATB24001
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Description:
    MycoLight™ Green JJ99 stain
  • Purity:
    95%

Specifications

About this item

MycoLight™ Green JJ99 stain is an excellent green-fluorescent nuclear and chromosome counterstain that is permeant to both prokaryotic and eukaryotic cell membranes.

  • Cell-Permeable
  • High DNA Affinity
  • Bright Fluorescence
Caution: For research use only.