6-ROXtra™ Fluorescence Reference Solution *25 µM for PCR Reactions*
Catalog # AATB398
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:5 mL
- Description:6-ROXtra™
- Purity:95%
Specifications
About this item
6-ROX is predominately used as a reference dye for performing PCR detections.
- Multiple bioconjugation applications
- 95% purity
Caution: For research use only.