FunXite™-1
Catalog # AATB22470
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 µl
- Description:FunXite™-1
- Purity:95%
Specifications
About this item
FunXite™-1 is chemically same molecule to FUN® 1 (FUN® 1 is the trademark of ThermoFisher).
- Bicolor Fluorescence For Live/Dead Cell Differentiation
- Cell-Permeable
Caution: For research use only.