ContactUsLinkComponent
Amplite® Fluorimetric Fluorescamine Protein Quantitation Kit *Blue Fluorescence*
Amplite® Fluorimetric Fluorescamine Protein Quantitation Kit *Blue Fluorescence*
Catalog # AATB11100
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Fluorimetric Fluorescamine Protein Quantitation Kit *Blue Fluorescence*
Catalog # AATB11100
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ fluorimetric fluorescamine protein quantitation kit *blue fluorescence*
  • Tests per kit:
    200 Tests
Specifications
About this item
Fluorescamine is intrinsically non-fluorescent but reacts rapidly with primary aliphatic amines, including those in peptides and proteins, to yield a blue-green-fluorescent derivative.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.