Amplite® Colorimetric Beta-Hydroxybutyrate (Ketone Body) Assay Kit
Catalog # AATB13830
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Amplite™ colorimetric beta-hydroxybutyrate (ketone body) assay kit
Specifications
About this item
Ketone bodies are produced by the liver and used peripherally as an energy source when blood glucose levels drop.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.