ContactUsLinkComponent
Gelite™ Green Nucleic Acid Gel Staining Kit
Catalog # AATB17589
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Gelite™ Green Nucleic Acid Gel Staining Kit
Catalog # AATB17589
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Gelite™ green nucleic acid gel staining kit
Specifications
About this item
Gelite™ Green is a sensitive fluorescent nucleic acid gel stain for detecting nucleic acids in agarose and polyacrylamide gels.
  • 2× more sensitive than etbr
  • Brighter signal
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.