Cell Navigator® F-Actin Labeling Kit *Green Fluorescence*
Catalog # AATB22661
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:500 Tests
- Description:Cell Navigator™ F-actin labelling kit *green fluorescence*
Specifications
About this item
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labelling sub-cellular organelles such as membranes, lysosomes, mitochondria, nuclei and etc.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.