Cell Navigator® Lysosome Staining Kit *Blue Fluorescence*
AATB22655
:
- Pk:500 Tests
- Description:Cell Navigator™ lysosome staining kit *blue fluorescence*
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labelling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
: For research use only.