Order Entry
Germany
ContactUsLinkComponent
Cell Explorer™ Live Cell Labeling Kit *Orange Fluorescence with 405 nm Excitation*
Cell Explorer™ Live Cell Labeling Kit *Orange Fluorescence with 405 nm Excitation*
Catalog # AATB22616
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Explorer™ Live Cell Labeling Kit *Orange Fluorescence with 405 nm Excitation*
Catalog # AATB22616
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Explorer™ live Cell labelling kit *orange fluorescence with 405 nm excitation*

Specifications

About this item

Our Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.

  • Contains all necessary components
  • Multiple research applications
  • Large Stokes shift
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.