Cell Explorer™ Live Cell Labeling Kit *Blue Fluorescence with 405 nm Excitation*
Catalog # AATB22614
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Explorer™ live cell labelling kit *blue fluorescence with 405 nm excitation*
Specifications
About this item
Our Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.