ContactUsLinkComponent
Amplite® Colorimetric Xanthine Oxidase Assay Kit
Amplite® Colorimetric Xanthine Oxidase Assay Kit
Catalog # AATB11307
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Colorimetric Xanthine Oxidase Assay Kit
Catalog # AATB11307
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric xanthine oxidase assay kit
Specifications
About this item
Xanthine oxidase (XO)is an enzyme that catalyzes the oxidation of hypoxanthine to xanthine and can further catalyse the oxidation of xanthine to uric acid.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.