Order Entry
Germany
ContactUsLinkComponent
Cell Meter™ 2-NBDG Glucose Uptake Assay Kit
Cell Meter™ 2-NBDG Glucose Uptake Assay Kit
Catalog # AATB23500
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ 2-NBDG Glucose Uptake Assay Kit
Catalog # AATB23500
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ 2-NBDG glucose uptake assay kit

Specifications

About this item

Glucose metabolism, a process which converts glucose into energy, is a primary source of energy supply in most organisms.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.