Cell Navigator® Fluorimetric Lipid Droplet Assay Kit *Red Fluorescence*
Catalog # AATB22735
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Navigator™ fluorimetric lipid droplet assay kit *red fluorescence*
Specifications
About this item
Lipid droplets, also referred to as lipid bodies, oil bodies or adiposomes, are lipid-rich cellular organelles that regulate the storage and hydrolysis of neutral lipids.
- Live cell labeling
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.