ContactUsLinkComponent
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
Catalog # AATB22476
Supplier:  AAT Bioquest
CAS Number:  
undefined
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
Catalog # AATB22476
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
Specifications
About this item
Differentiates live yeast with intact membranes will stain fluorescent green, and dead yeast with damaged membranes will stain fluorescent red.
  • Robust protocol
  • Live yeast will glow green
  • Dead yeast will glow red
The kit uses CFDA-AM and Propidium Iodide. CFDA-AM is a cell permeable esterase substrate. The acetoxymethyl (AM) group of the substrate will allow CFDA to penetrate cell membranes. Once inside the cells, lipophilic blocking and diacetate groups are cleaved by esterases, resulting in green fluorescence. In contrast, Propidium Iodide only stain the yeasts with damaged membranes resulting in red fluorescence.
Convenient way to test viability of yeasts.