Cell Meter™ Cell Adhesion Assay Kit
Catalog # AATB23010
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ cell adhesion assay kit
Specifications
About this item
The Cell Meter™ cell adhesion assay kit is a fast and sensitive assay for measuring cell-cell or cell-surface adhesion for a variety of cell types.
- Contains all necessary components
- Suitable for most common cell types
- pH-independent fluorescence
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.