Cell Meter™ Fluorimetric Phagocytosis Assay Kit *Red Fluorescence*
Catalog # AATB21225
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ fluorimetric phagocytosis assay kit *red fluorescence*
Specifications
About this item
Phagocytosis is defined as the cellular uptake of particles within a plasma-membrane envelope by a cell.
- Ready to use in suspension
- No trypan blue quenching required
- Simultaneous detection of live cells/phagocytosis
- Suitable for all major fluorescence platforms
Caution: For research use only.