Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Blue Fluorescence Excited at 405 nm*
Catalog # AATB22835
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ phosphatidylserine apoptosis assay kit *blue fluorescence excited at 405 nm*
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.